7-Bromoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR3616-1G
APOSOR3616-1GEA 128 EUR
APOSOR3616-1G
7-Bromoquinoline-3-carboxylic acid
7-Bromoquinoline-3-carboxylic acid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437554
CAS-Nummer: 892874-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR