tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate 95+%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 4-(2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)piperazine-1-carboxylate
OR361624-250MG
APOSOR361624-250MGEA
213
EUR
APOSOR361624-250MG
tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate 95+%
tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate
Formel:
C₂₃H₃₇BN₂O₅ Molecular Weight: 432,37 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827855 CAS-Nummer: 1310404-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...