1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester 95%

Lieferant: Apollo Scientific

OR361679-1G
APOSOR361679-1GEA 323 EUR
APOSOR361679-1G
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester 95%
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₅H₂₉BN₂O₃Si
Lagertemperatur: Raumtemperatur
MDL: MFCD09037502
CAS-Nummer: 903550-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR