4-Trimethylsilylphenylboronic acid, pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Trimethyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane

OR361680-5G
APOSOR361680-5GEA 197 EUR
APOSOR361680-5G
4-Trimethylsilylphenylboronic acid, pinacol ester 98%
4-Trimethylsilylphenylboronic acid, pinacol ester
Formel: C₁₅H₂₅BO₂Si
Molecular Weight: 276.254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037796
CAS-Nummer: 1186026-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR