Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate 95+%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-acetate
OR361684-250MG
APOSOR361684-250MGEA
30.6
EUR
APOSOR361684-250MG
Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate 95+%
Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate
Formel:
C₁₂H₁₉BN₂O₄ Molecular Weight: 266,101 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660239 CAS-Nummer: 959585-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...