2,6-Dichlor-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%

Lieferant: Apollo Scientific

Synonyme: 2-(3,5-Dichloro-4-hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dichloro-4-hydroxyphenylboronic Acid Pinacol Ester

OR361686-1G
APOSOR361686-1GEA 58 EUR
APOSOR361686-1G
2,6-Dichlor-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formel: C₁₂H₁₅BCl₂O₃
Molecular Weight: 288,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20526385
CAS-Nummer: 1003298-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR