6-Brom-4-chlor-2-methylchinolin

Lieferant: Apollo Scientific
Gefahr

OR3617-500MG
APOSOR3617-500MGEA 48 EUR
APOSOR3617-500MG
6-Brom-4-chlor-2-methylchinolin
6-Bromo-4-chloro-2-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,530 g/mol
Schmelzpunkt: 76…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272373
CAS-Nummer: 53364-85-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR