N,2-Dimethylbutan-1-amine hydrochloride ≥98%
Lieferant: Apollo Scientific
Synonyme:
Methyl(2-methylbutyl)amine hydrochloride
OR361722-100MG
APOSOR361722-100MGEA
56.7
EUR
APOSOR361722-100MG
N,2-Dimethylbutan-1-amine hydrochloride ≥98%
N,2-Dimethylbutan-1-amine hydrochloride
Formel:
C₆H₁₅N·ClH Molecular Weight: 137,651 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18483183 CAS-Nummer: 130985-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...