(4-(4-Amino-3-methylphenyl)piperidin-1-yl)(cyclopropyl)methanone ≥98%
Lieferant: Apollo Scientific
OR361726-250MG
APOSOR361726-250MGEA
748
EUR
APOSOR361726-250MG
(4-(4-Amino-3-methylphenyl)piperidin-1-yl)(cyclopropyl)methanone ≥98%
(4-(4-Amino-3-methylphenyl)piperidin-1-yl)(cyclopropyl)methanone
Formel:
C₁₆H₂₂N₂O Molecular Weight: 258,359 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30530548 CAS-Nummer: 1852497-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...