(5R)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
Lieferant: Apollo Scientific
OR361727-100MG
APOSOR361727-100MGEA
1540
EUR
APOSOR361727-100MG
(5R)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
(5R)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate
Formel:
C₁₁H₂₀BrNO₂ Molecular Weight: 278,186 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30728490 CAS-Nummer: 2096990-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...