(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%

Lieferant: Apollo Scientific

OR361728-100MG
APOSOR361728-100MGEA 1540 EUR
APOSOR361728-100MG
(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate
Formel: C₁₁H₂₀BrNO₂
Molecular Weight: 278,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30728491
CAS-Nummer: 2096990-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR