N,3-Dimethylbutan-2-amine ≥98%

Lieferant: Apollo Scientific

OR361732-1G
APOSOR361732-1GEA 854 EUR
APOSOR361732-1G
N,3-Dimethylbutan-2-amine ≥98%
N,3-Dimethylbutan-2-amine
Formel: C₆H₁₅N
Molecular Weight: 101,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09047478
CAS-Nummer: 34317-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR