Methyl 1-(Hydroxymethyl)cyclopropanecarboxylate ≥98%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-(hydroxymethyl)cyclopropanecarboxylate

OR361736-250MG
APOSOR361736-250MGEA 60.9 EUR
APOSOR361736-250MG
Methyl 1-(Hydroxymethyl)cyclopropanecarboxylate ≥98%
Methyl 1-(Hydroxymethyl)cyclopropanecarboxylate
Formel: C₆H₁₀O₃
Molecular Weight: 130,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036399
CAS-Nummer: 88157-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR