2-Chloro-1-(6-methoxypyridin-3-yl)ethanone ≥98%
Lieferant: Apollo Scientific
OR361738-100MG
APOSOR361738-100MGEA
434
EUR
APOSOR361738-100MG
2-Chloro-1-(6-methoxypyridin-3-yl)ethanone ≥98%
2-Chloro-1-(6-methoxypyridin-3-yl)ethanone
Formel:
C₈H₈ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20635182 CAS-Nummer: 136591-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...