2-(2-Amino-3-methylphenyl)propan-2-ol ≥98%
Lieferant: Apollo Scientific
OR361739-100MG
APOSOR361739-100MGEA
556
EUR
APOSOR361739-100MG
2-(2-Amino-3-methylphenyl)propan-2-ol ≥98%
2-(2-Amino-3-methylphenyl)propan-2-ol
Formel:
C₁₀H₁₅NO Molecular Weight: 165,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20695378 CAS-Nummer: 59689-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...