4-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR3624-250MG
APOSOR3624-250MGEA 66.7 EUR
APOSOR3624-250MG
4-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid 98%
4-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid
Formel: C₁₃H₁₉BN₂O₄
Molecular Weight: 278,11 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027219
CAS-Nummer: 913835-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR