3-Carboxy-4-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Borono-2-methoxybenzoic acid, 3-Carboxy-4-methoxybenzeneboronic acid
OR3625-250MG
APOSOR3625-250MGEA
21.9
EUR
APOSOR3625-250MG
3-Carboxy-4-methoxyphenylboronsäure 98%
3-Carboxy-4-methoxyphenylboronsäure
Formel:
C₈H₉BO₅ Schmelzpunkt: 176…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203322 CAS-Nummer: 913836-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...