4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-(2-Bromoethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR3628-5G
APOSOR3628-5GEA 217 EUR
APOSOR3628-5G
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester
Formel: C₁₄H₂₀BBrO₃
Molecular Weight: 327.03 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027285
CAS-Nummer: 913836-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR