D(-)-Isoleucin
Lieferant: Apollo Scientific
Synonyme:
(2R,3R)-(-)-α-Amino-β-methylvaleriansäure, (2R,3R)-(-)-2-Amino-3-methylpentansäure, (2R,3R)-(-)-Isoleucin, H-D-Ile-OH
OR36341-1G
OR36341-5G
APOSOR36341-1GEA
217
EUR
APOSOR36341-1G
APOSOR36341-5G
D(-)-Isoleucin
D(-)-Isoleucin
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131.17 g/mol Siedepunkt: 235 Schmelzpunkt: 265…274 °C Dichte: 1.224 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064221 CAS-Nummer: 319-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...