2-Chloro-7-methylquinoline-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR3638-250MG
APOSOR3638-250MGEA 21.9 EUR
APOSOR3638-250MG
2-Chloro-7-methylquinoline-3-carbaldehyde
2-Chloro-7-methylquinoline-3-carbaldehyde
Formel: C₁₁H₈ClNO
Molecular Weight: 205,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01365806
CAS-Nummer: 68236-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR