2,6-Dimethylquinoline-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR3640-250MG
APOSOR3640-250MGEA 36.4 EUR
APOSOR3640-250MG
2,6-Dimethylquinoline-3-carboxylic acid 95%
2,6-Dimethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06253921
CAS-Nummer: 610261-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR