Selenophene-2-boronic acid

Lieferant: Apollo Scientific
Gefahr

OR3645-1G
APOSOR3645-1GEA 848 EUR
APOSOR3645-1G
Selenophene-2-boronic acid
Selenophene-2-boronic acid
Formel: C₄H₅BO₂Se
Lagertemperatur: Raumtemperatur
MDL: MFCD01319053
CAS-Nummer: 35133-86-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR