8-Ethyl-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Ethyl-4-hydroxyquinaldine, 8-Ethyl-2-methylquinolin-4-ol

OR3646-250MG
APOSOR3646-250MGEA 128 EUR
APOSOR3646-250MG
8-Ethyl-4-hydroxy-2-methylquinoline
8-Ethyl-4-hydroxy-2-methylquinoline
Formel: C₁₂H₁₃NO
Molecular Weight: 187,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272396
CAS-Nummer: 63136-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR