2-Amino-6-brom-3-methylchinolin
Lieferant: Apollo Scientific
|
Achtung
|
OR3650-1G
APOSOR3650-1GEA
725
EUR
APOSOR3650-1G
2-Amino-6-brom-3-methylchinolin
2-Amino-6-bromo-3-methylquinoline
Formel:
C₁₀H₉BrN₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
203506-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...