7-Chlorindol
Lieferant: Apollo Scientific
|
Achtung
|
OR3666-1G
APOSOR3666-1GEA
34.8
EUR
APOSOR3666-1G
7-Chlorindol
7-Chlorindol
Formel:
C₈H₆ClN Molecular Weight: 151.6 g/mol Schmelzpunkt: 57…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321301 CAS-Nummer: 53924-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...