1-Acetyl-2-methyl-5-indolinesulphonoyl chloride

Lieferant: Apollo Scientific
Gefahr

OR3671-1G
APOSOR3671-1GEA 163 EUR
APOSOR3671-1G
1-Acetyl-2-methyl-5-indolinesulphonoyl chloride
1-Acetyl-2-methyl-5-indolinesulphonoyl chloride
Formel: C₁₁H₁₂ClNO₃S
Molecular Weight: 273,74 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07841689
CAS-Nummer: 841275-78-3
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR