3-Brom-4-chlor-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR3674-1G
APOSOR3674-1GEA
203
EUR
APOSOR3674-1G
3-Brom-4-chlor-1H-indazol
3-Bromo-4-chloro-1H-indazole
Formel:
C₇H₄BrClN₂ Schmelzpunkt: 233…234 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781582 CAS-Nummer: 885521-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...