3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR3675-100MG
APOSOR3675-100MGEA
24.8
EUR
APOSOR3675-100MG
3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine
3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine
Formel:
C₇H₃BrCl₂N₂ Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09607939 CAS-Nummer: 952183-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...