5-Chloro-2-pyridinethiol 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Chloropyridine-2-thiol

OR3682-250MG
APOSOR3682-250MGEA 24.8 EUR
APOSOR3682-250MG
5-Chloro-2-pyridinethiol 95%
5-Chloro-2-pyridinethiol
Formel: C₅H₄ClNS
Molecular Weight: 145,61 g/mol
Schmelzpunkt: 195…198 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00661468
CAS-Nummer: 40771-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR