7-Amino-1H-indazol 95%

Lieferant: Apollo Scientific
Achtung

OR3685-1G
APOSOR3685-1GEA 21.9 EUR
APOSOR3685-1G
7-Amino-1H-indazol 95%
7-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 157…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022790
CAS-Nummer: 21443-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR