4-(4-Methoxyphenyl)pyrimidin-2-amine

Lieferant: Apollo Scientific
Achtung

OR3689-100MG
APOSOR3689-100MGEA 21.9 EUR
APOSOR3689-100MG
4-(4-Methoxyphenyl)pyrimidin-2-amine
4-(4-Methoxyphenyl)pyrimidin-2-amine
Formel: C₁₁H₁₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00665912
CAS-Nummer: 99844-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR