1-Methyl-1H-indazol-6-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR3690-500MG
OR3690-1G
OR3690-5G
APOSOR3690-500MGEA
31.9
EUR
APOSOR3690-500MG
APOSOR3690-1G
APOSOR3690-5G
1-Methyl-1H-indazol-6-amine
1-Methyl-1H-indazol-6-amine
Formel:
C₈H₉N₃ Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00456076 CAS-Nummer: 74728-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...