2-(2-Bromopyridin-4-yl)-1-(pyridin-2-yl)ethanone ≥98%

Lieferant: Apollo Scientific

OR370003-1G
APOSOR370003-1GEA 739 EUR
APOSOR370003-1G
2-(2-Bromopyridin-4-yl)-1-(pyridin-2-yl)ethanone ≥98%
2-(2-Bromopyridin-4-yl)-1-(pyridin-2-yl)ethanone
Formel: C₁₂H₉BrN₂O
Molecular Weight: 277,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09031951
CAS-Nummer: 446852-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR