4-Acridinecarboxylic acid 98
Lieferant: Apollo Scientific
Synonyme:
Acridine-4-carboxylic acid
OR370004-100MG
OR370004-1G
OR370004-250MG
APOSOR370004-100MGEA
78.3
EUR
APOSOR370004-100MG
APOSOR370004-1G
APOSOR370004-250MG
4-Acridinecarboxylic acid 98
4-Acridinecarboxylic acid
Formel:
C₁₄H₉NO₂ Molecular Weight: 223.227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21340176 CAS-Nummer: 31327-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...