4-Acridinecarboxylic acid 98

Lieferant: Apollo Scientific

Synonyme: Acridine-4-carboxylic acid

OR370004-100MG OR370004-1G OR370004-250MG
APOSOR370004-100MGEA 78.3 EUR
APOSOR370004-100MG APOSOR370004-1G APOSOR370004-250MG
4-Acridinecarboxylic acid 98
4-Acridinecarboxylic acid
Formel: C₁₄H₉NO₂
Molecular Weight: 223.227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21340176
CAS-Nummer: 31327-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR