(3-Amino-4-(4-methylpiperidin-1-yl)phenyl)methanol ≥98%

Lieferant: Apollo Scientific

OR370005-100MG
APOSOR370005-100MGEA 967 EUR
APOSOR370005-100MG
(3-Amino-4-(4-methylpiperidin-1-yl)phenyl)methanol ≥98%
(3-Amino-4-(4-methylpiperidin-1-yl)phenyl)methanol
Formel: C₁₃H₂₀N₂O
Molecular Weight: 220,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15201763
CAS-Nummer: 1158544-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR