5-Bromo-1,4-dimethyl-1H-benzotriazole ≥98%
Lieferant: Apollo Scientific
OR370008-100MG
APOSOR370008-100MGEA
1620
EUR
APOSOR370008-100MG
5-Bromo-1,4-dimethyl-1H-benzotriazole ≥98%
5-Bromo-1,4-dimethyl-1H-benzotriazole
Formel:
C₈H₈BrN₃ Molecular Weight: 226,073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28969713 CAS-Nummer: 1799973-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...