1-(Imidazo[1,2-a]pyridin-6-yl)ethanone ≥98%

Lieferant: Apollo Scientific

OR370013-100MG OR370013-1G OR370013-250MG
APOSOR370013-100MGEA 336 EUR
APOSOR370013-100MG APOSOR370013-1G APOSOR370013-250MG
1-(Imidazo[1,2-a]pyridin-6-yl)ethanone ≥98%
1-(Imidazo[1,2-a]pyridin-6-yl)ethanone
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD10696290
CAS-Nummer: 944905-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR