Methyl 1-(4-bromophenyl)-3-oxocyclobutanecarboxylate ≥98%
Lieferant: Apollo Scientific
OR370015-100MG
APOSOR370015-100MGEA
799
EUR
APOSOR370015-100MG
Methyl 1-(4-bromophenyl)-3-oxocyclobutanecarboxylate ≥98%
Methyl 1-(4-bromophenyl)-3-oxocyclobutanecarboxylate
Formel:
C₁₂H₁₁BrO₃ Molecular Weight: 283,118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199278 CAS-Nummer: 1364663-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...