4-Bromo-6,7-dimethoxy-indan-1-one ≥98%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-6,7-dimethoxy-2,3-dihydro-1H-inden-1-one

OR370017-100MG
APOSOR370017-100MGEA 554 EUR
APOSOR370017-100MG
4-Bromo-6,7-dimethoxy-indan-1-one ≥98%
4-Bromo-6,7-dimethoxy-indan-1-one
Formel: C₁₁H₁₁BrO₃
Molecular Weight: 271,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00088977
CAS-Nummer: 18028-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR