5-Methoxyquinoline-7-carbonitrile ≥98%
Lieferant: Apollo Scientific
OR370023-100MG
APOSOR370023-100MGEA
351
EUR
APOSOR370023-100MG
5-Methoxyquinoline-7-carbonitrile ≥98%
5-Methoxyquinoline-7-carbonitrile
Formel:
C₁₁H₈N₂O Molecular Weight: 184,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11878501 CAS-Nummer: 1378483-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...