1-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥98%
Lieferant: Apollo Scientific
OR370025-1G
APOSOR370025-1GEA
333
EUR
APOSOR370025-1G
1-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile ≥98%
1-(tert-Butyl)-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile
Formel:
C₁₂H₁₃N₃ Molecular Weight: 199,252 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529068 CAS-Nummer: 269726-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...