7-Oxaspiro[3.5]nonane-1,3-dione ≥98%

Lieferant: Apollo Scientific

OR370030-100MG
APOSOR370030-100MGEA 840 EUR
APOSOR370030-100MG
7-Oxaspiro[3.5]nonane-1,3-dione ≥98%
7-Oxaspiro[3.5]nonane-1,3-dione
Formel: C₈H₁₀O₃
Molecular Weight: 154,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18642680
CAS-Nummer: 455264-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR