3-Chloro-1-methyl-1H-pyrazol-4-amine ≥98%

Lieferant: Apollo Scientific

OR370032-100MG
APOSOR370032-100MGEA 368 EUR
APOSOR370032-100MG
3-Chloro-1-methyl-1H-pyrazol-4-amine ≥98%
3-Chloro-1-methyl-1H-pyrazol-4-amine
Formel: C₄H₆ClN₃
Molecular Weight: 131,564 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19214410
CAS-Nummer: 150187-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR