O-(2-Methylbenzyl)hydroxylamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR370034-100MG
APOSOR370034-100MGEA 248 EUR
APOSOR370034-100MG
O-(2-Methylbenzyl)hydroxylamine hydrochloride ≥98%
O-(2-Methylbenzyl)hydroxylamine hydrochloride
Formel: C₈H₁₁NO·ClH
Molecular Weight: 173.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18253006
CAS-Nummer: 115777-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR