8-Hydroxycoumarin 98

Lieferant: Apollo Scientific

OR370035-250MG
APOSOR370035-250MGEA 179 EUR
APOSOR370035-250MG
8-Hydroxycoumarin 98
8-Hydroxycoumarin
Formel: C₉H₆O₃
Molecular Weight: 162.142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18450722
CAS-Nummer: 2442-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR