4-(2-Amino-5-bromo-3-pyridinyl)-2-methyl-3-butyn-2-ol 95%

Lieferant: Apollo Scientific

Synonyme: 4-(2-Amino-5-bromopyridin-3-yl)-2-methylbut-3-yn-2-ol

OR370040-100MG
APOSOR370040-100MGEA 1130 EUR
APOSOR370040-100MG
4-(2-Amino-5-bromo-3-pyridinyl)-2-methyl-3-butyn-2-ol 95%
4-(2-Amino-5-bromo-3-pyridinyl)-2-methyl-3-butyn-2-ol
Formel: C₁₀H₁₁BrN₂O
Molecular Weight: 255,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19689609
CAS-Nummer: 1262985-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR