5-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indol-1-one ≥98%

Lieferant: Apollo Scientific

OR370043-100G
APOSOR370043-100GEA 1270 EUR
APOSOR370043-100G
5-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indol-1-one ≥98%
5-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indol-1-one
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458716
CAS-Nummer: 122852-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR