4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol ≥98%
Lieferant: Apollo Scientific
OR370047-100MG
APOSOR370047-100MGEA
789
EUR
APOSOR370047-100MG
4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol ≥98%
4-(2-(Hydroxymethyl)phenyl)-1-methyl-3-phenylpiperidin-4-ol
Formel:
C₁₉H₂₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18207051 CAS-Nummer: 218288-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...