2-Benzyl-2,5-diazaspiro[3.4]octan-1-one ≥98%

Lieferant: Apollo Scientific

OR370049-100MG
APOSOR370049-100MGEA 968 EUR
APOSOR370049-100MG
2-Benzyl-2,5-diazaspiro[3.4]octan-1-one ≥98%
2-Benzyl-2,5-diazaspiro[3.4]octan-1-one
Formel: C₁₃H₁₆N₂O
Molecular Weight: 216,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18207189
CAS-Nummer: 1415562-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR